Learn More
TARGETMOL CHEMICALS INC ROCK-IN-5 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T677485MG
Encompass_Preferred
Also available in 10mg 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. ROCK-IN-5 (compound I-B-37) is a potent inhibitor of protein kinases including ROCK ERK GSK and AGC. ROCK-IN-5 exhibits the potential for cardiac proliferative and neurodegenerative diseases research. purity: 99%
Catalog No. 50-257-6458