Learn More
Medchemexpress LLC HY-119937 5mg Medchemexpress, ROCK inhibitor-2 CAS:1127308-52-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1199375MG
Encompass_Preferred
Medchemexpress, HY-119937 5mg ROCK inhibitor-2 CAS:1127308-52-4 ROCK inhibitor-2 is a selective dual ROCK1 and ROCK2 inhibitor with IC50s of 17 nM and 2 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9558