Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rock inhibitor-2 (compound 6) | 1127308-52-4 | 99.9% | 25 MG
SDP

Catalog No. 5000234651
Encompass_Preferred

ROCK inhibitor-2 is a selective small-molecule inhibitor of Rho-associated protein kinases ROCK1 and ROCK2 that reduces phosphorylation of MYPT1 (pMYPT1) and has reported antitumor activity. It is supplied as a high-purity solid for laboratory research.

  • Selective dual inhibitor of ROCK1 and ROCK2.
  • Reduces pMYPT1 phosphorylation; IC50 values: ROCK1 160 nM, ROCK2 21 nM, pMYPT1 75 nM.
  • High purity: 99.9% (analytical).
  • Molecular weight 332.40 g/mol; formula C21H20N2O2.
  • Soluble in DMSO (≈200 mg/mL) and formulations achieving ≥5 mg/mL for in vivo use.
  • Powder storage: -20°C (up to 3 years) or 4°C (up to 2 years).
  • For research use only; not for human or animal therapeutic use.

Catalog No. 50-002-34651 Supplier Medchemexpress LLC Supplier No. HY11993725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ROCK inhibitor-2 is a selective small-molecule inhibitor of Rho-associated protein kinases ROCK1 and ROCK2 that reduces phosphorylation of MYPT1 (pMYPT1) and has reported antitumor activity. It is supplied as a high-purity solid for laboratory research.

  • Selective dual inhibitor of ROCK1 and ROCK2.
  • Reduces pMYPT1 phosphorylation; IC50 values: ROCK1 160 nM, ROCK2 21 nM, pMYPT1 75 nM.
  • High purity: 99.9% (analytical).
  • Molecular weight 332.40 g/mol; formula C21H20N2O2.
  • Soluble in DMSO (≈200 mg/mL) and formulations achieving ≥5 mg/mL for in vivo use.
  • Powder storage: -20°C (up to 3 years) or 4°C (up to 2 years).
  • For research use only; not for human or animal therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.