Learn More
ATCC Rock Inhibitor Y27632
Catalog No. 502382173
Encompass
ROCK Inhibitor Y27632 can be used in increased primary human organoid formation efficiency and recovery from thawing and passaging. The ROCK Inhibitor Y27632 can be used in enhanced survival and maintenance of pluripotency of human embryonic stem cells (hESC), human induced pluripotent stem cells (hiPSC), and 3-D culture of organoids. Also, the inhibitor can be used in critical media components in conditional reprogramming of cells (CRC) culture technology.
Catalog No. 50-238-2173 Supplier ATCC Supplier No. ACS3030
 Restricted Item
Add to Cart
Add to Cart
ROCK Inhibitor Y27632 can be used in increased primary human organoid formation efficiency and recovery from thawing and passaging. The ROCK Inhibitor Y27632 can be used in enhanced survival and maintenance of pluripotency of human embryonic stem cells (hESC), human induced pluripotent stem cells (hiPSC), and 3-D culture of organoids. Also, the inhibitor can be used in critical media components in conditional reprogramming of cells (CRC) culture technology.