Learn More
Rock and Mineral Test Kit
SDPSCI_ED
Catalog No. S44872
Click to view available options
Product Type:
Rock and Mineral Test Kit
Catalog No. Product Type
S44872 Rock and Mineral Test Kit
1 options
Catalog No. S44872 Supplier Geoscience Industries Supplier No. S44872MN/10
Identify rock and mineral characteristics.
Includes:
  • Black and white ceramic streak plates
  • Glass plate
  • Nail
  • Penny
  • Magnet
  • Dropping bottle
  • Magnifying lens
  • Mohs Hardness Scale chart
Specifications
For Use With (Application) Identify rock and mineral characteristics
Product Type Rock and Mineral Test Kit
Includes Black and white ceramic streak plates, Glass plate, nail, penny, magnet, dropping bottle, magnifying lens, mohs hardness scale chart