Learn More
Selleck Chemical LLC Y-27632 2HCl 2mg 129830-38-2
SDP
Catalog No. NC0529385
Encompass_Preferred
Y-27632 2HCl is a selective ROCK1 (p160ROCK) inhibitor with Ki of 140 nM in a cell-free assay, exhibits >200-fold selectivity over other kinases, including PKC, cAMP-dependent protein kinase, MLCK and PAK. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0529385 Supplier Selleck Chemical LLC Supplier No. S10492MG
Y-27632 2HCl is a selective ROCK1 (p160ROCK) inhibitor with Ki of 140 nM in a cell-free assay, exhibits >200-fold selectivity over other kinases, including PKC, cAMP-dependent protein kinase, MLCK and PAK. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.