Learn More
Carnabio Usa Inc ROCK1/10UG/CATALYTIC
Catalog No. 507540972
Encompass
ROCK1, Catalytic domain, Wild type, Amino Acid 1-477, NP_005397.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0972 Supplier Carnabio Usa Inc Supplier No. 0110910
ROCK1, Catalytic domain, Wild type, Amino Acid 1-477, NP_005397.1, Expressed in Insect (sf21); 10microg