Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rock1 protein, human (Sf9, His) | >90.0% | 10UG
SDP

Catalog No. 5000176873
Encompass_Preferred

Recombinant human ROCK1 protein produced in Sf9 insect cells and supplied with an N-terminal 8xHis tag. Supplied as a 10 μg aliquot in buffered solution (50 mM HEPES, 200 mM NaCl, 20% glycerol, 1 mM DTT, pH 7.5), the protein is intended for biochemical assays, activity studies, and antibody validation. It is shipped on dry ice and should be stored at -80°C; avoid repeated freeze-thaw cycles.

  • Recombinant human ROCK1 kinase expressed in Sf9 insect cells.
  • N-terminal 8xHis tag facilitates purification and detection.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Approximate molecular weight 50 kDa.
  • Supplied in 50 mM HEPES, 200 mM NaCl, 20% glycerol, 1 mM DTT, pH 7.5.
  • Aliquoted as 10 μg for small-scale assays and validation experiments.
  • Shipped on dry ice; store at -80°C and avoid repeated freeze-thaw cycles.

Catalog No. 50-001-76873 Supplier Medchemexpress LLC Supplier No. HYP70177010UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human ROCK1 protein produced in Sf9 insect cells and supplied with an N-terminal 8xHis tag. Supplied as a 10 μg aliquot in buffered solution (50 mM HEPES, 200 mM NaCl, 20% glycerol, 1 mM DTT, pH 7.5), the protein is intended for biochemical assays, activity studies, and antibody validation. It is shipped on dry ice and should be stored at -80°C; avoid repeated freeze-thaw cycles.

  • Recombinant human ROCK1 kinase expressed in Sf9 insect cells.
  • N-terminal 8xHis tag facilitates purification and detection.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Approximate molecular weight 50 kDa.
  • Supplied in 50 mM HEPES, 200 mM NaCl, 20% glycerol, 1 mM DTT, pH 7.5.
  • Aliquoted as 10 μg for small-scale assays and validation experiments.
  • Shipped on dry ice; store at -80°C and avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.