Learn More
Carnabio Usa Inc ROCK2/5UG/CATALYTIC
Supplier:  Carnabio Usa Inc 011105
Encompass
ROCK2, Catalytic domain, Wild type, Amino Acid 1-553, NP_004841.2, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0611