Learn More
Sino Biological Recombinant Rat ROCK2 Protein (GST Tag), Active
Catalog No. 5000083955
Encompass_Preferred
Recombinant rat ROCK2 (5-554) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-000-83955 Supplier Sino Biological Supplier No. R11A11G10
Recombinant rat ROCK2 (5-554) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.