Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rock2-in-2 | 1995065-79-6 | MFCD31692339 | 98.1% | 360.39 g/mol | C18H12N6OS | 100 MG
SDP

Catalog No. 5000244918
Encompass_Preferred

ROCK2-IN-2 is a selective inhibitor of Rho-associated protein kinase 2 (ROCK2) described in patent US20180093978A1 (Compound A-30). It is supplied as a solid research compound with reported ROCK2 IC50 <1 μM and is intended for in vitro and cellular studies.

  • Selective ROCK2 inhibition with IC50 <1 μM
  • High purity: 98.05%
  • Molecular formula: C18H12N6OS; molecular weight: 360.39 g/mol
  • Appearance: light yellow to green yellow solid
  • Soluble in DMSO (100 mg/mL); ultrasonic assistance recommended
  • Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months)

Catalog No. 50-002-44918 Supplier Medchemexpress LLC Supplier No. HY103620100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ROCK2-IN-2 is a selective inhibitor of Rho-associated protein kinase 2 (ROCK2) described in patent US20180093978A1 (Compound A-30). It is supplied as a solid research compound with reported ROCK2 IC50 <1 μM and is intended for in vitro and cellular studies.

  • Selective ROCK2 inhibition with IC50 <1 μM
  • High purity: 98.05%
  • Molecular formula: C18H12N6OS; molecular weight: 360.39 g/mol
  • Appearance: light yellow to green yellow solid
  • Soluble in DMSO (100 mg/mL); ultrasonic assistance recommended
  • Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.