Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ROCK2, GST Tag Recombinant Protein

Catalog No. PV3759
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV3759 10 μg
PR7116B 100 μg
PV4048 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PV3759 Supplier Invitrogen™ Supplier No. PV3759
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Catalytic.

ROCK2, the predominant ROCK isoform in skeletal muscle, is an important regulator downstream of Rho GTPases. ROCK2 regulates cytokinesis, smooth muscle contraction, the formation of actin stress fibers and focal adhesions, and the activation of the c-fos serum response element. This protein, which is an isozyme of ROCK1 is a target for the small GTPase Rho.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75116
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 0.02% Triton X-100, 50% glycerol, 2mM DTT, 0.5mM EDTA, 150mM NaCl and no preservative; pH 7.5
Gene ID (Entrez) 9475
Molecular Weight (g/mol) 90.8 kDa
Name Human ROCK2, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias B230113H15Rik; KIAA0619; mKIAA0619; p150 ROK-alpha; p164 ROCK-2; Rho associated coiled-coil containing protein kinase 2; Rho kinase 2; RhoA - binding serine/threosine kinase alpha (ROK - alpha); rhoA-binding kinase 2; Rho-associated coiled-coil containing protein kinase 2; Rho-associated coiled-coil forming kinage 2; Rho-associated coiled-coil forming kinase 2; rho-associated protein kinase 2; Rho-associated, coiled-coil-containing protein kinase 2; rho-associated, coiled-coil-containing protein kinase II; Rho-kinase; ROCK II; Rock2; Rock2m; rockii; ROCK-II; ROK; ROKalpha
Common Name ROCK2
Gene Symbol ROCK2
Kinase Family DMPK Kinase Family
Protein Length 1-552
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 1-552
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Kinase Group Ser⁄Thr Kinases: ACG Group
Shipping Condition Dry Ice
System Compatibility LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate
Tag Position N-Terminal
Target Entry ROCK2
Target Gene Rho-associated coiled-coil containing protein kinase 2
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Catalytic
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.