Learn More
Electron Microscopy Sciences Rocker Tape Sheets, 12" x 12"
SDP
Catalog No. 5033747
Encompass_Preferred
Double-sided adhesive film for laboratory container. No more fumbling with clamps. nUse with rockers, shakers, incubators. or nutators. Easily rock and roll up to 300rpm and beyond. Re-usable based on load capacity and vessel number. Keeps on rockin' even in the prescence of most solvents and caustic agents.
Catalog No. 50-337-47 Supplier Electron Microscopy Sciences Supplier No. 7751220
Double-sided adhesive film for laboratory container. No more fumbling with clamps. nUse with rockers, shakers, incubators. or nutators. Easily rock and roll up to 300rpm and beyond. Re-usable based on load capacity and vessel number. Keeps on rockin' even in the prescence of most solvents and caustic agents.