Learn More
Sterlitech Corporation Rocker 300 Vacuum Pump, 110V/60Hz
SDP
Catalog No. 501952291
Encompass_Preferred
Horse Power: 1/8, Flow Rate (lpm): 23Vacuum Pump Applications:* Filtration for Microbiology Test* Air Sampling* Vacuum Pad Hold-Down* Suspended Solid Test* Vacuum Filtration
Catalog No. 50-195-2291 Supplier Sterlitech Corporation Supplier No. 16730011
Horse Power: 1/8, Flow Rate (lpm): 23Vacuum Pump Applications:* Filtration for Microbiology Test* Air Sampling* Vacuum Pad Hold-Down* Suspended Solid Test* Vacuum Filtration