Learn More
New Star Environmental Inc ROCKER 300 PUMP/110VLT
Catalog No. NC0895629
Encompass_Preferred
Oil Free Vacuum Pump. Rocker 300 Vacuum Pump, 21 liters/minute, AC110V, 60Hz; Features: •No air pollution, maintenance free. •Rocker series pumps are driven by Piston, without the need of lubricant, regular oil changes and maintenance; with no oil pollution.
Catalog No. NC0895629 Supplier New Star Environmental Inc Supplier No. 16730011
Oil Free Vacuum Pump. Rocker 300 Vacuum Pump, 21 liters/minute, AC110V, 60Hz; Features: •No air pollution, maintenance free. •Rocker series pumps are driven by Piston, without the need of lubricant, regular oil changes and maintenance; with no oil pollution.