Learn More
Sterlitech Corporation Rocker 300C-VF12 Vacuum Filtration System, includes Rocker 300C Pump, VF 12 Filtration Set and 1m Silicone Tubing (AC110V/60Hz)
SDP
Catalog No. 501952297
Encompass_Preferred
Application: Filtration System for Solvent.Membrane disc filter diameter: 47mmSterlitech offers kits to enable fast, easy set-up for performing a wide variety of vacuum filtration applications.
Catalog No. 50-195-2297 Supplier Sterlitech Corporation Supplier No. 18930211
Application: Filtration System for Solvent.Membrane disc filter diameter: 47mmSterlitech offers kits to enable fast, easy set-up for performing a wide variety of vacuum filtration applications.