Learn More
Sterlitech Corporation Rocker 400 Vacuum Pump, 110V/60Hz
SDP
Catalog No. NC1450738
Encompass_Preferred
Flow Rate (lpm):37 Horsepower:1/6 HPPower: 110V/60Hz Tubing: 5/16" hose barb weight: 11.5 lbs
Catalog No. NC1450738 Supplier Sterlitech Corporation Supplier No. 16740011
Flow Rate (lpm):37 Horsepower:1/6 HPPower: 110V/60Hz Tubing: 5/16" hose barb weight: 11.5 lbs