Learn More
Bellco Glass, Inc. Rocker Complete, 100V-25x25 Tray
SDP
Catalog No. 501946114
Encompass_Preferred
The Bellco Rocker Platforms provide a gentle uniform rocking motion, required for even coverage of media over mono-layer cell cultures, thereby maximizing cellular health and viability. Viral suspensions introduced onto a mono-layer of cells will yield higher virus titers. Additionally, rocker platforms are ideal for performing staining and destaining of gel and filter assays. They are engineered to operate between 0 to 12 oscillations per minute. Units are delivered with either a 25 x 25 cm or 51 x 51 cm tray. Additional interchangeable trays are also available.. MADE IN THE USA--Speed range: 0 to 12 oscillations per minute--Platform tilt: 7 fixed
Catalog No. 50-194-6114 Supplier Bellco Glass, Inc. Supplier No. 774010100
The Bellco Rocker Platforms provide a gentle uniform rocking motion, required for even coverage of media over mono-layer cell cultures, thereby maximizing cellular health and viability. Viral suspensions introduced onto a mono-layer of cells will yield higher virus titers. Additionally, rocker platforms are ideal for performing staining and destaining of gel and filter assays. They are engineered to operate between 0 to 12 oscillations per minute. Units are delivered with either a 25 x 25 cm or 51 x 51 cm tray. Additional interchangeable trays are also available.. MADE IN THE USA--Speed range: 0 to 12 oscillations per minute--Platform tilt: 7 fixed