Learn More
Chemglass Life Sciences Digital Rocker, 120V, Base Only, Versa-Rock
SDP
Catalog No. 50143341
Encompass
Digital Rocker, 120V, Base Only, Versa-Rock. • 24V Brushless Low Maintenance DC Motor and Speed Controller• Digital Speed Display• Locking Speed Control Knob• Peg and Grommet Tray Design• 2-Year WarrantyThe Versa-Rock is a digital slow-speed rocker for general laboratory cell culture work. The low profile design incorporates a rocking platform that accepts a variety of optional trays. The Peg and Grommet style trays make changing convenient and interchangeable with other similar trays. Trays with liners sold separately.NOTE: Trays are not supplied with base and must be purchased separately.
Catalog No. 50-143-341 Supplier Chemglass Life Sciences Supplier No. CLS4033100
Digital Rocker, 120V, Base Only, Versa-Rock. • 24V Brushless Low Maintenance DC Motor and Speed Controller• Digital Speed Display• Locking Speed Control Knob• Peg and Grommet Tray Design• 2-Year WarrantyThe Versa-Rock is a digital slow-speed rocker for general laboratory cell culture work. The low profile design incorporates a rocking platform that accepts a variety of optional trays. The Peg and Grommet style trays make changing convenient and interchangeable with other similar trays. Trays with liners sold separately.NOTE: Trays are not supplied with base and must be purchased separately.