Learn More
Thomas Scientific ProBlot Rocker 25 with single platform (26 x 20 cm), 120V
SDP
Catalog No. 501457693
Encompass
The Labnet Economy Rockers, ProBlot™ 25 and 25XL high capacity bench rockers, are basic bench rocking platforms that have been designed for a long life and trouble-free service. Staining, hybridization, washing, etc. Compact size, low profile Extremely quiet Double platform option expands usable space Bench rocker speed is adjustable to provide the gentle motion required for staining fragile gels as well as the more vigorous action necessary for washing blots. The rocking movement is fixed at ±7°. The platform surfaces are covered with nonslip rubber pads to prevent objects from moving during operation. Elastic tie downs are available to secure in place bags, boxes, tubes and other containers. The platform assemblies pivot on permanently lubricated ball bearings for quiet, maintenance-free operation. The ProBlot™ 25 platform measures 28.9 x 20.3 cm, while the ProBlot™ 25XL platform is slightly larger at 30 x 30 cm. For increasing the work area without sacri
Catalog No. 50-145-7693 Supplier Thomas Scientific Supplier No. 1189F96
The Labnet Economy Rockers, ProBlot™ 25 and 25XL high capacity bench rockers, are basic bench rocking platforms that have been designed for a long life and trouble-free service. Staining, hybridization, washing, etc. Compact size, low profile Extremely quiet Double platform option expands usable space Bench rocker speed is adjustable to provide the gentle motion required for staining fragile gels as well as the more vigorous action necessary for washing blots. The rocking movement is fixed at ±7°. The platform surfaces are covered with nonslip rubber pads to prevent objects from moving during operation. Elastic tie downs are available to secure in place bags, boxes, tubes and other containers. The platform assemblies pivot on permanently lubricated ball bearings for quiet, maintenance-free operation. The ProBlot™ 25 platform measures 28.9 x 20.3 cm, while the ProBlot™ 25XL platform is slightly larger at 30 x 30 cm. For increasing the work area without sacri