Learn More
Grainger Rocker Switch
Catalog No. 502748746
Encompass_Preferred
The Columbus McKinnon Rocker Switch 4K1R-157F17 is a heavy duty rocker switch designed for industrial applications. It has a 15 amp, 480VAC rating and is UL/CSA certified. The switch has quick connect terminals and is supplied with a legend plate. It can be mounted to panels up to 1/4" thick using the included hardware. This rocker switch is made in the USA by Columbus McKinnon, a leading manufacturer of material handling products. With its high amperage rating and rugged construction, this switch is well-suited for controlling motors, conveyors, pumps and other industrial equipment.
Catalog No. 50-274-8746 Supplier Grainger Supplier No. 157F17
The Columbus McKinnon Rocker Switch 4K1R-157F17 is a heavy duty rocker switch designed for industrial applications. It has a 15 amp, 480VAC rating and is UL/CSA certified. The switch has quick connect terminals and is supplied with a legend plate. It can be mounted to panels up to 1/4" thick using the included hardware. This rocker switch is made in the USA by Columbus McKinnon, a leading manufacturer of material handling products. With its high amperage rating and rugged construction, this switch is well-suited for controlling motors, conveyors, pumps and other industrial equipment.