Learn More
Thomas Scientific Rocking Platform
SDP
Catalog No. 501460092
Encompass
Excellent temperature uniformity and accuracy Interior outlets for powering small equipment Drip pans to catch and clean spills Wide, variable speed range Broad temperature range Adjustable rotisserie The Hybridization oven holds six large bottles and up to twelve small bottles. Rotisserie is unique in that it will hold 50 mL tubes, eliminating the need to purchase a separate one for processing strips and small blots. Featuring a corrosion resistant stainless steel interior and a grounded interior electrical outlet for powering small accessory equipment. Includes two large bottles and one pack of mesh. A rotisserie for 35 mm bottles and a drip pan is installed in each oven chamber. Compatible with all standard hybridization ovens Heavy-walled glass construction Teflon seal Hybridization bottles, mesh and bottle rack are compatible with all standard hybridization ovens (Hybaid, Boekel, UVP, etc.). The bottles’heavy-walled, borosilicate glass construction blocks most beta radi
Catalog No. 50-146-0092 Supplier Thomas Scientific Supplier No. 1159Z88
Excellent temperature uniformity and accuracy Interior outlets for powering small equipment Drip pans to catch and clean spills Wide, variable speed range Broad temperature range Adjustable rotisserie The Hybridization oven holds six large bottles and up to twelve small bottles. Rotisserie is unique in that it will hold 50 mL tubes, eliminating the need to purchase a separate one for processing strips and small blots. Featuring a corrosion resistant stainless steel interior and a grounded interior electrical outlet for powering small accessory equipment. Includes two large bottles and one pack of mesh. A rotisserie for 35 mm bottles and a drip pan is installed in each oven chamber. Compatible with all standard hybridization ovens Heavy-walled glass construction Teflon seal Hybridization bottles, mesh and bottle rack are compatible with all standard hybridization ovens (Hybaid, Boekel, UVP, etc.). The bottles’heavy-walled, borosilicate glass construction blocks most beta radi