Learn More
Cole-Parmer Labnet Rocking Platform Shaker, Double, 115 VAC
Supplier:  Cole-Parmer 5140112
Encompass_Preferred
Platform rockers are designed for durability and ease of use. . Simple analog controls for both speed and time make give the use precise control over rocking parameters. Adjust the tilt angle up to 20° with knob on side of rocker.
Available in both single and dual platform models and includes a ribbed no-slip rubber mat to keep containers in place.
Catalog No. NC1745673