Learn More
Research Products International Corp Double Platform Rocking Shaker, 12 x 16 Inches
SDP
Catalog No. 50550223
Encompass
This rocking shaker provides the optimum agitation for blotting membranes or staining gels. Electronic speed control accurately maintains set speed from 4 to 160 motions per minute. Produces a uniform and gentle wave pattern that washes over and under membranes. Adjustable platform with locking tilt can be set from 0 to 20° arc. Can be placed inside an incubator up to 65°C. Five year warranty.
Catalog No. 50-550-223 Supplier Research Products International Corp Supplier No. 195552
This rocking shaker provides the optimum agitation for blotting membranes or staining gels. Electronic speed control accurately maintains set speed from 4 to 160 motions per minute. Produces a uniform and gentle wave pattern that washes over and under membranes. Adjustable platform with locking tilt can be set from 0 to 20° arc. Can be placed inside an incubator up to 65°C. Five year warranty.