Learn More
Strem, An Ascensus Company CAS# 2009020-38-4. 10g. Methanesulfonato(2-(di-t-butylphosphino)-3-methoxy-6-methyl-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II), min. 98% [RockPhos Palladacycle Gen. 3]. MFCD27952542
SDP
Supplier:  Strem, An Ascensus Company 46033510G
Encompass
CAS# 2009020-38-4. 10g. Methanesulfonato(2-(di-t-butylphosphino)-3-methoxy-6-methyl-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II), min. 98% [RockPhos Palladacycle Gen. 3]. MFCD27952542. Molecular Weight: 838.43. Molecular Formula: C44H62NO4PPdS. Color/form: brown pwdr. Strem# 46-0335. http://www.strem.com/catalog/v/46-0335/
Catalog No. 50-201-6741