Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Rockphos Pd G3

Catalog No. 7739055G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
800 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-390-55G 500 mg
77-390-5500MG 100 mg
77-390-5100MG 800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 77-390-55G Supplier Sigma Aldrich Supplier No. 7739055G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Rockphos Pd G3

Specifications

CAS 2009020-38-4
Melting Point 220°C to 250°C
Molecular Formula C44H62NO4PPdS
Linear Formula C44H62NO4PPdS
MDL Number MFCD27952542
Quantity 500 mg
Synonym RockPhos-G3-palladacycle; [(2-Di-tert-butylphosphino-3-methoxy-6-methyl-2',4',6'-triisopropyl-1,1'-biphenyl)-2-(2-aminobiphenyl)]palladium(II) methanesulfonate
Molecular Weight (g/mol) 838.43 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.