Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Rocks Learning Center
SCI_ED

Catalog No. S24169 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S24169 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24169 Supplier NewPath Learning™ Supplier No. 246879
Add to Cart
Add to Cart

Engaging, ready-to-use learning centers for review of standards-based science skills.

  • For learning essential grade-appropriate science skills
  • Designed for teacher-directed instruction or independent student group learning
  • Materials sufficient for up to four students
  • Allows to use at classroom, after-school programs, or at home
  • Includes one Curriculum Mastery Game featuring:
    1. One set of 30 illustrated self-checking question cards
    2. One laminated double-sided game board and game pieces for up to 4 players
  • Four Visual Learning Guides focused on same topic
  • Packed in zippered storage pouch
  • For Grades 6, 7, 8, High School

Specifications

Grade Level Intermediate - Middle School, High School
Grade 6 to 12
Description Rocks Learning Center
Product Type Learning Center
Includes
  • One set of 30 illustrated self-checking question cards
  • One laminated double-sided game board and game pieces for up to 4 players
  • Four Visual Learning Guides focused on same topic
Certifications Academic standard: California, Florida, National, New York, Texas
For Use With (Application) For teacher-directed instruction or independent student group learning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.