Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rocuronium bromide | 119302-91-9 | MFCD00867768 | 98.0% | 609.68 g/mol | C32H53BrN2O4 | 10 MG
SDP

Catalog No. 5000298423
Encompass_Preferred

Rocuronium bromide is an aminosteroid, non-depolarizing neuromuscular blocking agent supplied as a white to off-white solid for laboratory research use. It is commonly used to study nicotinic acetylcholine receptor interactions and neuromuscular transmission in vitro. This material is intended for research purposes only and is not for clinical, diagnostic, or therapeutic use.

  • Purity approximately 98.0%.
  • White to off-white solid appearance.
  • Molecular weight 609.68 g/mol.
  • Soluble in DMSO and water (≈100 mg/mL); ultrasonic may be required.
  • Store solid at 4°C; store solutions at -80°C (up to 6 months) or -20°C (up to 1 month) when sealed.
  • Provided in milligram quantities suitable for laboratory assays.

Catalog No. 50-002-98423 Supplier Medchemexpress LLC Supplier No. HY1744010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rocuronium bromide is an aminosteroid, non-depolarizing neuromuscular blocking agent supplied as a white to off-white solid for laboratory research use. It is commonly used to study nicotinic acetylcholine receptor interactions and neuromuscular transmission in vitro. This material is intended for research purposes only and is not for clinical, diagnostic, or therapeutic use.

  • Purity approximately 98.0%.
  • White to off-white solid appearance.
  • Molecular weight 609.68 g/mol.
  • Soluble in DMSO and water (≈100 mg/mL); ultrasonic may be required.
  • Store solid at 4°C; store solutions at -80°C (up to 6 months) or -20°C (up to 1 month) when sealed.
  • Provided in milligram quantities suitable for laboratory assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.