Learn More
Apexbio Technology LLC Rocuronium Bromide 119302-91-9 25mg
SDP
Catalog No. 502240782
Encompass_Preferred
Rocuronium Bromide (CAS 119302-91-9) is an aminosteroid compound that functions as a non-depolarizing neuromuscular blocking agent It acts by competitively antagonizing nicotinic acetylcholine receptors at the neuromuscular junction thereby inhibiting acetylcholine-induced depolarization and subsequent muscle contraction In research settings Rocuronium Bromide is utilized to investigate neuromuscular transmission and to model pharmacological blockade of skeletal muscle activity supporting studies in anesthesia muscle physiology and synaptic transmission mechanisms
This item is not returnable. View return policy
Catalog No. 50-224-0782 Supplier Apexbio Technology LLC Supplier No. A136625
Rocuronium Bromide (CAS 119302-91-9) is an aminosteroid compound that functions as a non-depolarizing neuromuscular blocking agent It acts by competitively antagonizing nicotinic acetylcholine receptors at the neuromuscular junction thereby inhibiting acetylcholine-induced depolarization and subsequent muscle contraction In research settings Rocuronium Bromide is utilized to investigate neuromuscular transmission and to model pharmacological blockade of skeletal muscle activity supporting studies in anesthesia muscle physiology and synaptic transmission mechanisms