Learn More
Medchemexpress LLC Rocuronium Bromide (ORG 9426 Bromide) | 119302-91-9 | 98.0% | 609.68 | 5 MG
SDP
Catalog No. 5000386691
Encompass_Preferred
Rocuronium Bromide is an aminosteroid non-depolarizing neuromuscular blocker or muscle relaxant used in modern anaesthesia, to facilitate endotracheal intubation and to provide skeletal muscle relaxation during surgery or mechanical ventilation. It is intended for research use only.
  • Aminosteroid non-depolarizing neuromuscular blocker
  • Muscle relaxant
  • Used in modern anaesthesia
  • Facilitates endotracheal intubation
  • Provides skeletal muscle relaxation during surgery or mechanical ventilation
  • For research use only
Catalog No. 50-003-86691 Supplier Medchemexpress LLC Supplier No. HY174405MG
Rocuronium Bromide is an aminosteroid non-depolarizing neuromuscular blocker or muscle relaxant used in modern anaesthesia, to facilitate endotracheal intubation and to provide skeletal muscle relaxation during surgery or mechanical ventilation. It is intended for research use only.
  • Aminosteroid non-depolarizing neuromuscular blocker
  • Muscle relaxant
  • Used in modern anaesthesia
  • Facilitates endotracheal intubation
  • Provides skeletal muscle relaxation during surgery or mechanical ventilation
  • For research use only