Learn More
Cayman Chemical Rocuronium (bromide), Purity: ≥95%, CAS: 119302-91-9, 50 mg
SDP
Catalog No. 502995184
Encompass_Preferred
A non-depolarizing muscle relaxant; selectively induces neuromuscular blockade in cats (ED50s = 153, 185, 1,329, and 4,119 μg/kg for tibialis, soleus, and vagus muscles, and nictitating membranes, respectively; has a more rapid onset time than vecuronium
Catalog No. 50-299-5184 Supplier Cayman Chemical Supplier No. 2369850 MG
A non-depolarizing muscle relaxant; selectively induces neuromuscular blockade in cats (ED50s = 153, 185, 1,329, and 4,119 μg/kg for tibialis, soleus, and vagus muscles, and nictitating membranes, respectively; has a more rapid onset time than vecuronium