Learn More
Chemglass Life Sciences Clamp Connector Rod End for 1/2 in Support Tubes
SDP
Catalog No. NC2745617
Encompass_Preferred
Clamp connector rod end designed to join 1/2 in (12.7 mm) support tubes or rods, providing a cross-block connection for laboratory support assemblies.
  • Provides cross-block connection for 1/2 in support tubes.
  • Compatible with 1/2 in (12.7 mm) O.D. support tubes or rods.
  • Requires 5/32 in Allen wrench for socket head cap screw.
  • Sold individually.
Catalog No. NC2745617 Supplier Chemglass Life Sciences Supplier No. CG926301
Clamp connector rod end designed to join 1/2 in (12.7 mm) support tubes or rods, providing a cross-block connection for laboratory support assemblies.
  • Provides cross-block connection for 1/2 in support tubes.
  • Compatible with 1/2 in (12.7 mm) O.D. support tubes or rods.
  • Requires 5/32 in Allen wrench for socket head cap screw.
  • Sold individually.