Learn More
Grainger Rod Stock Brass 1/16 in x 3 ft PK10
Catalog No. 502764536
Encompass_Preferred
The 260 Brass Rod, 1/4" Outside Diameter, is a 1/4" diameter brass rod stock that is 36" long. It is made from 260 brass alloy which contains 70% copper and 30% zinc, and is known for having good formability and corrosion resistance. This rod has a smooth surface finish and meets ASTM B16 standards for brass rod. With a tensile strength of 55,000 psi, it can be used for various machining, fabrication and mechanical applications. The rod weighs approximately 0.75 lbs.
Catalog No. 50-276-4536 Supplier Grainger Supplier No. 16NH91
The 260 Brass Rod, 1/4" Outside Diameter, is a 1/4" diameter brass rod stock that is 36" long. It is made from 260 brass alloy which contains 70% copper and 30% zinc, and is known for having good formability and corrosion resistance. This rod has a smooth surface finish and meets ASTM B16 standards for brass rod. With a tensile strength of 55,000 psi, it can be used for various machining, fabrication and mechanical applications. The rod weighs approximately 0.75 lbs.