Learn More
Grainger Rod Stock Brass 1/4 in x 3 ft PK4
Catalog No. 502764078
Encompass_Preferred
The 260 Brass Rod is a 1/4 inch diameter rod made of 260 brass alloy. It is 36 inches long and has a smooth surface finish. The 260 brass alloy contains 70% copper and 30% zinc, which provides good ductility and corrosion resistance. This rod stock meets ASTM B16 standards and has many applications including shafts, pins, rivets, and more. With a tensile strength of 50,000 psi, this 360 brass rod offers good machinability and formability. Overall, it is a versatile 360 brass round rod well-suited for fabrication and machining.
Catalog No. 50-276-4078 Supplier Grainger Supplier No. 16NH95
The 260 Brass Rod is a 1/4 inch diameter rod made of 260 brass alloy. It is 36 inches long and has a smooth surface finish. The 260 brass alloy contains 70% copper and 30% zinc, which provides good ductility and corrosion resistance. This rod stock meets ASTM B16 standards and has many applications including shafts, pins, rivets, and more. With a tensile strength of 50,000 psi, this 360 brass rod offers good machinability and formability. Overall, it is a versatile 360 brass round rod well-suited for fabrication and machining.