Learn More
Grainger Rod Stock Brass 3/8 in x 3 ft PK3
Catalog No. 502764080
Encompass_Preferred
The 260 Brass Rod 3/8 in Outside Diameter x 36 in Length is a solid brass rod with a 3/8 inch outside diameter and a length of 36 inches. It is made of 260 brass alloy which contains 70% copper and 30% zinc, and meets ASTM B16 standards. This corrosion resistant rod has excellent machinability and ductility. It can be used for shafts, bearings, bushings, and other precision machined parts and components. With its 360 Brinell hardness, the rod offers good wear resistance while still being easy to machine. Overall, this 3 foot long brass rod provides versatile material for machining, fabrication and metalworking applications.
Catalog No. 50-276-4080 Supplier Grainger Supplier No. 16NH97
The 260 Brass Rod 3/8 in Outside Diameter x 36 in Length is a solid brass rod with a 3/8 inch outside diameter and a length of 36 inches. It is made of 260 brass alloy which contains 70% copper and 30% zinc, and meets ASTM B16 standards. This corrosion resistant rod has excellent machinability and ductility. It can be used for shafts, bearings, bushings, and other precision machined parts and components. With its 360 Brinell hardness, the rod offers good wear resistance while still being easy to machine. Overall, this 3 foot long brass rod provides versatile material for machining, fabrication and metalworking applications.