Learn More
Grainger Rod Stock Brass 5/16 in x 3 ft PK3
Catalog No. 502764079
Encompass_Preferred
The Grainger Brass Rod Stock is 3 feet long with a diameter of 5/16 inches. It is made from 360 brass which has excellent corrosion resistance and is easy to machine. The rod has a smooth surface finish and meets ASTM B16 standards. With a tensile strength of 48,000 psi, this 360 brass rod stock can be used for applications like bushings, spacers, and prototypes. It comes in 3 foot lengths and can be cut to size.
Catalog No. 50-276-4079 Supplier Grainger Supplier No. 16NH96
The Grainger Brass Rod Stock is 3 feet long with a diameter of 5/16 inches. It is made from 360 brass which has excellent corrosion resistance and is easy to machine. The rod has a smooth surface finish and meets ASTM B16 standards. With a tensile strength of 48,000 psi, this 360 brass rod stock can be used for applications like bushings, spacers, and prototypes. It comes in 3 foot lengths and can be cut to size.