Learn More
Grainger Rod Stock Brass 5/32 in x 3 ft PK5
Catalog No. 502764527
Encompass_Preferred
The 260 Brass Rod is a 5/32 inch diameter rod made of 260 brass alloy. It has excellent machinability and can be used for shafts, bearings, bushings and other precision parts. The rod has a smooth finish and tight tolerances. It is available in 12 foot lengths and can be cut to size. The 260 brass alloy contains 70% copper and 30% zinc, which gives it good corrosion resistance. This brass rod product is made in the USA by Grainger, a leading industrial supplier. It meets ASTM B16 standards for brass rod.
Catalog No. 50-276-4527 Supplier Grainger Supplier No. 16NH94
The 260 Brass Rod is a 5/32 inch diameter rod made of 260 brass alloy. It has excellent machinability and can be used for shafts, bearings, bushings and other precision parts. The rod has a smooth finish and tight tolerances. It is available in 12 foot lengths and can be cut to size. The 260 brass alloy contains 70% copper and 30% zinc, which gives it good corrosion resistance. This brass rod product is made in the USA by Grainger, a leading industrial supplier. It meets ASTM B16 standards for brass rod.