Learn More
Grainger Durham Manufacturing Open Portable Truck, 2-Shelf, 3,000 lb. Capacity, 60 in. x 30 in., 1 ea.
Catalog No. 502790364
Encompass_Preferred
This heavy-duty open portable truck is a reliable solution for organizing and transporting materials in garages, warehouses, and shipping departments. Featuring a durable welded 14-gauge construction, this unit is designed to handle demanding industrial environments with an overall capacity of 3,000 lbs.
  • Solid construction with a textured powder coat finish
  • Includes one fixed shelf and one base shelf for versatile storage
  • Equipped with four 6 in. x 2 in. phenolic casters (two rigid, two swivel) for easy mobility
  • Flat bar cage design on three sides for enhanced stability
  • Ships fully assembled for immediate use
  • Load capacity of 1,500 lbs. per shelf
Catalog No. 50-279-0364 Supplier Grainger Supplier No. 16C285
This heavy-duty open portable truck is a reliable solution for organizing and transporting materials in garages, warehouses, and shipping departments. Featuring a durable welded 14-gauge construction, this unit is designed to handle demanding industrial environments with an overall capacity of 3,000 lbs.
  • Solid construction with a textured powder coat finish
  • Includes one fixed shelf and one base shelf for versatile storage
  • Equipped with four 6 in. x 2 in. phenolic casters (two rigid, two swivel) for easy mobility
  • Flat bar cage design on three sides for enhanced stability
  • Ships fully assembled for immediate use
  • Load capacity of 1,500 lbs. per shelf