Learn More
Grainger Durham Manufacturing Bulk Stock Cart, 3-Shelf, 3000 lb Capacity, 14 Gauge Steel, 1 ea
Catalog No. 502788799
Encompass_Preferred
This bulk stock cart is designed for heavy-duty material handling with a 3,000 lb. total load capacity. Constructed from 14-gauge steel with a textured powder coat finish, this three-shelf unit provides a durable solution for organizing and transporting inventory in industrial environments.
  • 3,000 lb. overall capacity with 1,000 lb. capacity per shelf.
  • Includes two sloped shelves and one flat bottom shelf.
  • Equipped with 6 in. x 2 in. phenolic casters, featuring two rigid and two swivel configurations for mobility.
  • Welded construction with 14-gauge steel for long-term durability.
  • Tubular push handle for ease of maneuverability.
  • Flush lip style on solid shelves.
Catalog No. 50-278-8799 Supplier Grainger Supplier No. 16C281
This bulk stock cart is designed for heavy-duty material handling with a 3,000 lb. total load capacity. Constructed from 14-gauge steel with a textured powder coat finish, this three-shelf unit provides a durable solution for organizing and transporting inventory in industrial environments.
  • 3,000 lb. overall capacity with 1,000 lb. capacity per shelf.
  • Includes two sloped shelves and one flat bottom shelf.
  • Equipped with 6 in. x 2 in. phenolic casters, featuring two rigid and two swivel configurations for mobility.
  • Welded construction with 14-gauge steel for long-term durability.
  • Tubular push handle for ease of maneuverability.
  • Flush lip style on solid shelves.