Learn More
BD RODAC™ SNAP LID Sabouraud Dextrose Agar (SDA) with Lecithin and Polysorbate 80 (LP80)
Catalog No. B215282 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
Packaging Type:
Sterile Double Wrapping Plate
VHP Resistant Triple Wrapping Isolator Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
B215282 100/Pk. Sterile Double Wrapping Plate
B215270 10/Pk. VHP Resistant Triple Wrapping Isolator Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B215282 Supplier BD Supplier No. 215282
This item is not returnable. View return policy
The BD RODAC SNAP LID is a RODAC contact plate equipped with a unique SNAP LID locking mechanism to provide additional security during sampling and transport from a controlled environment to the laboratory.
The RODAC SNAP LID has a latch on the base of the plate that engages the ridge on the lid of the plate to lock the plate; that:
  • Withstands an accidental drop from the height of a bench-top workspace
  • Ensures the integrity of the sample
  • Prevents the dish from opening unintentionally
  • Protects media from accidental cross contamination
  • Improves stability during stacking and storage
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Form Liquid
Manufacturing Origin United States
Media Type Sabouraud Dextrose Agar with Lecithin and Polysorbate 80
Sterility Sterile
Includes Rodac™ snap lid
Packaging Type Sterile Double Wrapping Plate
Quantity 100/Pk.