Learn More
Heidolph™ Rodaviss Connection Accessories
SDP
Catalog No. 05403943 Shop All Heidolph North America Products
Change view
Click to view available options
Type:
Rodaviss Connecting Cap
Rodaviss Connecting Set
Connection Type:
B14/23
B24/29
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Connection Type
05-403-943 Rodaviss Connecting Set B24/29
05-403-785 Rodaviss Connecting Cap B14/23
05-403-781 Rodaviss Connecting Set B14/23
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05-403-943 Supplier Heidolph™ Supplier No. 015880510
Add to Cart
Add to Cart
Heidolph™ Rodaviss Connection Accessories optimize glass connections to ensure they remain stable
  • Suitable for work performed under vacuum and pressure
  • The connections remain absolutely stable and can be easily separated again due to components that are technically matched to each other
  • The connection comprises cone and socket as well as cap, safeguarding and sealing ring
  • Can be used up to a temperature of 200°C
  • Minimum breakage risk due to anti-jamming system
  • Replaceable with ground joints in accordance with DIN 12 249
Specifications
Connection Type B24/29
Type Rodaviss Connecting Set
For Use With (Application) Optimizing Glass Connections
For Use With (Equipment) Carousel 6 Plus Reaction System
Quantity 10/Pk.
Temperature (Metric) Range Up to 200°C
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov