Learn More
MSC BASF Rodent Bait, Soft Bait, 8 lb Pail, Cholecalciferol
Catalog No. 502835961
Encompass
This rodent bait, featuring cholecalciferol as its active ingredient, is designed for efficient and rapid control of rodent infestations. Its unique stop-feed action ensures rodents consume only a lethal dose, reducing bait waste. The formulation is highly palatable and effective against anticoagulant-resistant rodents, offering reliable performance even in challenging environmental conditions.
  • Controls entire infestations in seven days.
  • Lowers labor and material costs.
  • Shorter baiting programs require less bait.
  • Active ingredient causes stop-feed action, meaning rodents eat only a lethal dose.
  • Effective against anticoagulant-resistant rodents.
  • Proven palatability even when other food sources are available.
  • Durable in tough conditions, resisting extreme temperatures.
  • Low risk of secondary, non-target poisoning to birds and mammals.
Catalog No. 50-283-5961 Supplier MSC Supplier No. 74357484
This rodent bait, featuring cholecalciferol as its active ingredient, is designed for efficient and rapid control of rodent infestations. Its unique stop-feed action ensures rodents consume only a lethal dose, reducing bait waste. The formulation is highly palatable and effective against anticoagulant-resistant rodents, offering reliable performance even in challenging environmental conditions.
  • Controls entire infestations in seven days.
  • Lowers labor and material costs.
  • Shorter baiting programs require less bait.
  • Active ingredient causes stop-feed action, meaning rodents eat only a lethal dose.
  • Effective against anticoagulant-resistant rodents.
  • Proven palatability even when other food sources are available.
  • Durable in tough conditions, resisting extreme temperatures.
  • Low risk of secondary, non-target poisoning to birds and mammals.