Learn More
Biocare Medical, LLC Rodent Block M - 25 ml
SDP
Catalog No. 5083263
Encompass_Preferred
Rodent Block M is an immunohistochemical reagent designed for blocking endogenous mouse IgG and non-specific background in mouse tissues and red blood cells. Rodent Block M can be used with Mouse-on-Mouse,cells. Rodent Block M can be used with Mouse-on-Mouse, on-Mouse detection systems. After the application of Rodent Block M, the desired primary antibody can be applied. It can
Catalog No. 50-832-63 Supplier Biocare Medical, LLC Supplier No. RBM961H
Rodent Block M is an immunohistochemical reagent designed for blocking endogenous mouse IgG and non-specific background in mouse tissues and red blood cells. Rodent Block M can be used with Mouse-on-Mouse,cells. Rodent Block M can be used with Mouse-on-Mouse, on-Mouse detection systems. After the application of Rodent Block M, the desired primary antibody can be applied. It can