Learn More
Biocare Medical, LLC Rodent Block R - 100 ml
SDP
Catalog No. 5083267
Encompass_Preferred
Rodent Block R is an immunohistochemical reagent designed for blocking endogenous rat IgG and non-specific background in rat tissues and red blood cells. After the application of Rodent Block R, a mouse monoclonal antibody can be applied. It can also be used on an automated staining system.
Catalog No. 50-832-67 Supplier Biocare Medical, LLC Supplier No. RBR962L
Rodent Block R is an immunohistochemical reagent designed for blocking endogenous rat IgG and non-specific background in rat tissues and red blood cells. After the application of Rodent Block R, a mouse monoclonal antibody can be applied. It can also be used on an automated staining system.