Learn More
Electron Microscopy Sciences Acrylic Rodent Brain Matrix Adult Mouse, Coronal
SDP
Catalog No. 5029263
Encompass_Preferred
Low cost acrylic matrices are precisely machined and highly polished to ensure reproducible results. These matrices allow the investigator to slice either coronal (perpendicular to center line) or sagittal (parallel to center line) sections through the brain at 1 mm intervals.
Type: Coronal
Catalog No. 50-292-63 Supplier Electron Microscopy Sciences Supplier No. 69080C
Low cost acrylic matrices are precisely machined and highly polished to ensure reproducible results. These matrices allow the investigator to slice either coronal (perpendicular to center line) or sagittal (parallel to center line) sections through the brain at 1 mm intervals.
Type: Coronal