Learn More
Electron Microscopy Sciences Acrylic Rodent Brain Matrix Rat, 175-300g, Sagittal
SDP
Supplier:  Electron Microscopy Sciences 69081S
Encompass_Preferred
Low cost acrylic matrices are precisely machined and highly polished to ensure reproducible results. These matrices allow the investigator to slice either coronal (perpendicular to center line) or sagittal (parallel to center line) sections through the brain at 1 mm intervals.
Size: 175-300g
Type: Sagittal
Catalog No. 50-292-62