Learn More
Electron Microscopy Sciences Acrylic Rodent Brain Matrix Rat, 175-300g, Coronal
SDP
Catalog No. 5029260
Encompass_Preferred
Low cost acrylic matrices are precisely machined and highly polished to ensure reproducible results. These matrices allow the investigator to slice either coronal (perpendicular to center line) or sagittal (parallel to center line) sections through the brain at 1 mm intervals.
Size: 175-300g
Type: Coronal
Catalog No. 50-292-60 Supplier Electron Microscopy Sciences Supplier No. 69081C
Low cost acrylic matrices are precisely machined and highly polished to ensure reproducible results. These matrices allow the investigator to slice either coronal (perpendicular to center line) or sagittal (parallel to center line) sections through the brain at 1 mm intervals.
Size: 175-300g
Type: Coronal