Learn More
Alpco Ltd Chemi Rodent Insulin ELISA
SDP
Supplier:  Alpco Ltd 80INSMRCH01
Encompass_Preferred
The ALPCO Chemi Rodent Insulin ELISA measures the concentration of insulin protein products in mouse I, mouse II, rat I and rat II proinsulin genes. The kit has the performance characteristics and flexibility necessary to confidently measure up to 37 samples in duplicate. For Research Use Only. Not for Use in Diagnostic Procedures.
  • Low sample volume of 5 µL
  • Dynamic Range of 0.1-150 ng/mL
  • Superior sensitivity of 0.089 ng/mL
Catalog No. NC0544405