Learn More
Braintree Scientific Rodent Jacket, Mouse, 25-30g
SDP
Catalog No. 501955705
Encompass
Rodent Jackets. Manufactured from breathable, latex-free power net mesh in a saddle-style format. Designed for use with rodent tethers and available in a range of sizes to ensure a secure yet comfortable fit. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5705 Supplier Braintree Scientific Supplier No. RJM02
Rodent Jackets. Manufactured from breathable, latex-free power net mesh in a saddle-style format. Designed for use with rodent tethers and available in a range of sizes to ensure a secure yet comfortable fit. **Sold for research purposes only and not for human use**