Learn More
DRE MEDICAL GROUP INC RODENT NOSE CONE-FMASKS XSML
Catalog No. NC3714584
Encompass_Preferred
Rodent Nose Cone/Face Masks: for Mice and Rats XSmall. Weanling mice: X-small nose cone is 17mm diameter, Rodent face masks are made from clear acrylic and fit all non-rebreathing systems.
Catalog No. NC3714584 Supplier DRE MEDICAL GROUP INC Supplier No. P05096XS
Rodent Nose Cone/Face Masks: for Mice and Rats XSmall. Weanling mice: X-small nose cone is 17mm diameter, Rodent face masks are made from clear acrylic and fit all non-rebreathing systems.